RHC-80267 ≥98% (by TLC)

Toimittaja: ENZO LIFE SCIENCES

Synonyms: Cyclohexanone O-((6-((((cyclohexylideneamino)oxy)carbonyl)amino)hexyl)carbamoyl) oxime, RHC 80267

BMLST3000010 BMLST3000050
ENZOBMLST3000010EA 86.9 EUR
ENZOBMLST3000010 ENZOBMLST3000050
RHC-80267 ≥98% (by TLC)
RHC-80267

Diacylglycerol lipase inhibitor


Diacylglycerol (DAG) is an important cellular source of arachidonate which may be released by the action of DAG lipase. RHC-80267 selectively inhibits DAG lipase activity in canine platelets (IC50=4 µM), bovine adrenal chromaffin cells, human adrenal glomerulosa cells, rat thyroid lobes and pancreatic minilobules.

Kaava: C₂₀H₃₄N₄O₄
MW: 394.516 g/mol
MDL numero: MFCD00210844
CAS-numero: 83654-05-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=942123b42e618500dd5ab8e6d284a1f42ff4590f&vid=2977ec9c9bfd1f80c771ec5f48dbb57f836036ba&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR