3-O-Methyl-Sphingomyelin
Toimittaja: ENZO LIFE SCIENCES
BMLSL2250001 BMLSL2250005
ENZOBMLSL2250001EA 181 EUR
ENZOBMLSL2250001 ENZOBMLSL2250005
3-O-Methyl-Sphingomyelin
3-O-Methyl-Sphingomyelin
Sphingomyelinase inhibitor
3-O-Methyl-Sphingomyelin is a novel selective inhibitor of neutral sphingomyelinase, (Ki=37 µM). It is cell-permeable and does not inhibit acid sphingomyelinase.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35c492026c711ab3a8874e4d4363b5570cae0f08&vid=dbfff375eb1c0766d445244bd0dc3a03186dcfeb&prid=web&clid=SAPDAM