Phytosphingosine ≥98%

Toimittaja: ENZO LIFE SCIENCES

BMLSL1500005 BMLSL1500025
ENZOBMLSL1500005EA 89.1 EUR
ENZOBMLSL1500005 ENZOBMLSL1500025
Phytosphingosine ≥98%
Phytosphingosine

Ultra-pure


An endogenous sphingosine occurring in Saccharomyces cerevisiae. Inhibits FMLP-induced oxidant release by human neutrophils.

Kaava: C₁₈H₃₉NO₃
MDL numero: MFCD00079232
CAS-numero: 554-62-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1f34c340112e09534474ea51457c5230afe77a9&vid=adff813f3b7498f64cbe691600d8cbfd2eba7ce6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR