Human recombinant Shp-1 (from E. coli)
Toimittaja: ENZO LIFE SCIENCES
BMLSE3340020
ENZOBMLSE3340020EA 516 EUR
ENZOBMLSE3340020
Human recombinant Shp-1 (from E. coli)
Proteiinit
Produced in E. coli. Full length human Shp-1 (aa 1-595).