Anti-Y Mouse Monoclonal Antibody (Biotin) [clone: ]

Toimittaja: ENZO LIFE SCIENCES
BMLSA2430100
ENZOBMLSA2430100EA 321 EUR
ENZOBMLSA2430100
Anti-Y Mouse Monoclonal Antibody (Biotin) [clone: ]
Vasta-aineet
Host: Mouse, Isotype: IgG2b

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry

Recognizes phosphotyrosine. Species independent. No cross-reactivity with phosphoserine or phosphothreonine.

Type: Primary
Antigen: Y
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Many Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR