Leukotriene E4 ≥98% (by HPLC)

Toimittaja: ENZO LIFE SCIENCES
Vaara

BMLLE0040050 BMLLE0041000
ENZOBMLLE0040050EA 325 EUR
ENZOBMLLE0040050 ENZOBMLLE0041000
Leukotriene E4 ≥98% (by HPLC)
Leukotriene E4

Prostanoid receptor ligand.


Less potent than LTD4 in smooth muscle contractile activity and mucus secretion in airways. Reduces myocardial contractility and coronary blood flow. Induces long lasting hypotension and airway hyperresponsiveness. Partial agonist at LTD4 receptors. LTE4 solely activates a CysLT1 receptor. Biosynthesis from LTD4 by a dipeptidase.

Kaava: C₂₃H₃₇NO₅S
Storage Temperature: Ultrasyväjääpakastin (–80°C)
CAS-numero: 75715-89-8
UN: 1992
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=620a9bd70d9e5c33f1342742ad3c988167d9015e&vid=18cfadcf0d357f83801f60d1b1d06b698bcf2a10&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Concentration 50 µg/ml
Formulation Solution in methanol/ammonium acetate buffer, 70:30, pH 5.6.
Purity ≥98% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR