BML-210 ≥98% (by TLC)

Toimittaja: ENZO LIFE SCIENCES

Synonyms: CAY10433

BMLGR3300001 BMLGR3300005
ENZOBMLGR3300001EA 110 EUR
ENZOBMLGR3300001 ENZOBMLGR3300005
BML-210 ≥98% (by TLC)
BML-210

HDAC inhibitor developed at Enzo Life Sciences. Inhibits HDAC activity in HeLa nuclear extracts (IC50=5-10µM).


  • HDAC inhibitor


Tan solid.


Soluble in DMSO (25mg/ml) or 100% ethanol (10mg/ml, warm)

Kaava: C₂₀H₂₅N₃O₂
Storage Temperature: Pakastin
MDL numero: MFCD08062139
CAS-numero: 537034-17-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f4e13ae38c8b295361e899011e590c3638a9940&vid=68f17a50e559d4c67e71d6d33fe5a1a6d2f6cd60&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR