AM 580 ≥98% (by TLC)

Toimittaja: ENZO LIFE SCIENCES

Synonyms: NSC 608001, CD336, 4-​​[[(5,​​6,​​7,​​8-​​tetrahydro-​​5,​​5,​​8,​​8-​​tetramethyl-​​2-​​naphthalenyl)​carbonyl]​amino]​-​benzoic acid, AM580, 4-(5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalene-2-carboxamido)benzoic acid, RO 40-6055

BMLGR1040005 BMLGR1040025
ENZOBMLGR1040005EA 165 EUR
ENZOBMLGR1040005 ENZOBMLGR1040025
AM 580 ≥98% (by TLC)
AM 580

Retinoic acid analog which acts as a selective RARα agonist with no affinity for RXRs. Induces differentiation in leukemia cell lines and inhibits angiogenesis in chick embryos.


  • Retinoic acid receptor ligand


White to off-white solid.


Soluble in DMSO (25mg/ml) and ethanol (10mg/ml).

Kaava: C₂₂H₂₅NO₃
Storage Temperature: Pakastin
MDL numero: MFCD00673916
CAS-numero: 102121-60-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbca291264cfaa35adf2204220afbb5b6c286be4&vid=c6d9ccedeac2847b0a7ee3738d0bcc622bd166b9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR