N,N'-Di(2-hydroxybenzyl)ethylenediamine-N,N'-diacetic acid monohydrochloride hydrate (HBED) ≥95% (by NMR)

Toimittaja: ENZO LIFE SCIENCES

BMLFR1130250
ENZOBMLFR1130250EA 299 EUR
ENZOBMLFR1130250
N,N'-Di(2-hydroxybenzyl)ethylenediamine-N,N'-diacetic acid monohydrochloride hydrate (HBED) ≥95% (by NMR)
N,N'-Di(2-hydroxybenzyl)ethylenediamine-N,N'-diacetic acid monohydrochloride hydrate (HBED)

Potent iron chelator


Iron chelator. Inhibits ferric ion catalyzed formation of hydroxyl radicals. A very potent ligand for the chelation of ferric iron in vivo (active in rat models but inactive in primate models). Inhibits neutrophil apoptosis. An important tool for probing the involvement of hydroxyl radicals in cellular processes.

Kaava: C₂₀H₂₄N₂O₆·HCl·xH₂O
MDL numero: MFCD04113603
CAS-numero: 35369-53-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1e2abc3d9e036f7f61433914d61a1eddd63be08&vid=e3e27274415f72a625b202ac649979621c0c0675&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR