AG-879 ≥99%

Toimittaja: ENZO LIFE SCIENCES

BMLEI2580005 BMLEI2580025
ENZOBMLEI2580005EA 110 EUR
ENZOBMLEI2580005 ENZOBMLEI2580025
AG-879 ≥99%
AG-879

NGF receptor inhibitor


AG-879, a member of the tyrphostin family of tyrosine kinase inhibitors, inhibits NGF receptor (pp140c-trk) autophosphorylation selectively with no inhibition of EGF or PDGF receptor phosphorylation (IC50=10 µM). AG-879 inhibits NGF-induced neurite outgrowth in PC12 cells.

Kaava: C₁₈H₂₄N₂OS
MDL numero: MFCD00236450
CAS-numero: 148741-30-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d257a7ae48208c22e4c80565a4e694b0580a5773&vid=79ab55768f87e7ed097ba988b8efb6528283efcb&prid=web&clid=SAPDAM

Order Now

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR