Leukotriene B4-dimethylamide ≥98% (by HPLC)

Toimittaja: ENZO LIFE SCIENCES

BMLEI1020050
ENZOBMLEI1020050EA 451 EUR
ENZOBMLEI1020050
Leukotriene B4-dimethylamide ≥98% (by HPLC)
Leukotriene B4-dimethylamide

Leukotriene B4 antagonist. Inhibits binding of [3H]-LTB4 to guinea pig lung membranes (Ki=0.13 µM). Antagonizes the degranulation response to LTB4 in rabbit PMNL (Kd=0.2 µM) acting, most likely, via receptor desensitization. Partial agonist at higher concentrations (5 µM).


  • Prostanoid receptor ligand


Liquid.

Kaava: C₂₂H₃₇NO₃
Storage Temperature: Ultrasyväjääpakastin (–80°C)
CAS-numero: 83024-92-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cb1c8f85aa990dd8dd0d58e4d174bb54710a165&vid=6fc92a230e038f18dd39931041a91e00645a8d59&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR