25-Hydroxyvitamin D3 ≥98% (by HPLC), white to off-white solid

Toimittaja: ENZO LIFE SCIENCES
Vaara

BMLDM1000001 BMLDM1000005
ENZOBMLDM1000001EA 71.5 EUR
ENZOBMLDM1000001 ENZOBMLDM1000005
25-Hydroxyvitamin D3 ≥98% (by HPLC), white to off-white solid
25-Hydroxyvitamin D3

Precursor to 1α,25-dihydroxyvitamin D₃


Intermediate in the biosynthesis of 1alpha,25-dihydroxyvitamin D3. Major circulating metabolite of vitamin D3.


Soluble in 100% ethanol.

Kaava: C₂₇H₄₄O₂
Storage Temperature: Pakastin
MDL numero: MFCD00867077
CAS-numero: 19356-17-3
UN: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3ec23887b3d8e238fd5ef1c6f5940df673eb623&vid=ce4a390b2c87e0097d88cb0406310e7e5e60a035&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Identity Determined by IR
Purity (HPLC) ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR