AM-630 ≥98% (by TLC)

Toimittaja: ENZO LIFE SCIENCES
Varoitus

BMLCR1070010
ENZOBMLCR1070010EA 229 EUR
ENZOBMLCR1070010
AM-630 ≥98% (by TLC)
AM-630

Cannabinoid receptor ligand.


Acts as a cannabinoid receptor antagonist in mouse brain, vas deferens and guinea pig brain, but an agonist in guinea pig ileum. Acts as an inverse agonist on cloned human CB1 receptors.


Soluble in DMSO or dimethyl formamide.

Kaava: C₂₃H₂₅IN₂O₃
Storage Temperature: Huoneenlämpö
CAS-numero: 164178-33-0
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0e29dfb661953f8540046e5ac87e33ba8813373&vid=288119327b6e805735e1233899c0a1803645ef56&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Purity ≥98% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR