Sp-8-Br-cGMPS ≥98% (by HPLC)

Toimittaja: ENZO LIFE SCIENCES

BMLCN2170500
ENZOBMLCN2170500EA 66 EUR
ENZOBMLCN2170500
Sp-8-Br-cGMPS ≥98% (by HPLC)
Sp-8-Br-cGMPS

cGMP agonist.


Cell-permeable, phosphodiesterase-resistant cGMP agonist. This analog activates PKA and PKG with similar potency. Much more lipophilic and membrane permeant as compared to cyclic GMP.


Soluble in water (25 mg/ml).

Kaava: C₁₀H₁₀BrN₅O₆PS·Na
Storage Temperature: Pakastin
CAS-numero: 153660-03-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f76c6e0dead2356b9d00fd8d2957857c1454c600&vid=4cf237dfcced17380436ce0e6ec51d3e21d2a4bd&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Purity ≥98% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR