Dibutyryl-cAMP sodium salt ≥98% (by HPLC)

Toimittaja: ENZO LIFE SCIENCES

BMLCN1250030 BMLCN1250100
ENZOBMLCN1250030EA 88 EUR
ENZOBMLCN1250030 ENZOBMLCN1250100
Dibutyryl-cAMP sodium salt ≥98% (by HPLC)
Dibutyryl-cAMP sodium salt

PKA activator.


A cell-permeable cAMP analog which preferentially activates cAMP-dependent protein kinase. The products releaes butyrate due to intracellular and extracellular esterase action. Butyrate was shown to have distinct biological effects.


Soluble in water (100 mg/ml), DMSO (200 mM) or 96% ethanol (25 mM).

Kaava: C₁₈H₂₃N₅O₈P·Na
Storage Temperature: Pakastin
MDL numero: MFCD00005843
CAS-numero: 16980-89-5
EINECS: 241-059-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01751242474e209b481659378043656b9f7cae4e&vid=174f765272dc5542a1079f769a8709dfa0a865b7&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Purity ≥98% (HPLC)
Identity Determined by UV spectrum


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR