Matrix Metalloproteinase-8 (MMP-8) fluorometric drug discovery kit
Toimittaja: ENZO LIFE SCIENCES
BMLAK4150001
ENZOBMLAK4150001EA 680 EUR
ENZOBMLAK4150001
Matrix Metalloproteinase-8 (MMP-8) fluorometric drug discovery kit
Bio-assayt Entsymaattiset pikatestit
A QuantiZyme™ Assay System. The MMP-8 Fluorometric (also known as fluorimetric) Drug Discovery Kit is a complete assay system designed to screen inhibitors of matrix metalloproteinase-8 (MMP-8, neutrophil collagenase, collagenase-2) using a quenched fluorogenic peptide: OmniMMP™ fluorogenic substrate Mca-Pro-Leu-Gly-Leu-Dpa-Ala-Arg-NH2. Mca fluorescence is quenched by the Dpa group until cleavage by MMPs at the Gly-Leu bond separates the two moieties
Toimitustiedot: Kit contains MMP-8 Enzyme (human) (recombinant), Substrate, Calibration Standard, Prototypic Control Inhibitor (NNGH3), Assay Buffer, 96-well White Microplate and a detailed instruction booklet.