CD40L Soluble (Human) ELISA kit
Toimittaja: ENZO LIFE SCIENCES
ALX850311KI01
ENZOALX850311KI01EA 1050 EUR
ENZOALX850311KI01
CD40L Soluble (Human) ELISA kit
Bio-assayt ELISA-kitit
For the quantitative determination of soluble human CD40L (CD154) in cell culture supernatants, serum and other body fluids.
  • Assay Time: 3 hours + 10 minutes