sVEGFR-1 (Human) ELISA kit
Toimittaja: ENZO LIFE SCIENCES
ALX850264KI01
ENZOALX850264KI01EA 866 EUR
ENZOALX850264KI01
sVEGFR-1 (Human) ELISA kit
Bio-assayt ELISA-kitit
For the quantitative determination of human sVEGFR-1 in cell culture supernatants, serum and other body fluids.
  • Assay Time: 3,5 hours