Anti-BIMS/EL/L Rat Monoclonal Antibody

Toimittaja: ENZO LIFE SCIENCES
ALX804351C100
ENZOALX804351C100EA 533 EUR
ENZOALX804351C100
Anti-BIMS/EL/L Rat Monoclonal Antibody
Vasta-aineet
Host: Rat, Isotype: IgG2a

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse and monkey BimL, BimEL and BimS.

Type: Primary
Antigen: BIMS/EL/L
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR