Human;Mouse recombinant Thy-1 (RLE) (mouse):Fc
Toimittaja: ENZO LIFE SCIENCES
ALX522097C050
ENZOALX522097C050EA 592 EUR
ENZOALX522097C050
Human;Mouse recombinant Thy-1 (RLE) (mouse):Fc
Proteiinit
Produced in HEK 293 cells. The extracellular domain of mouse Thy-1 (aa 20-131) is fused to the Fc portion of human IgG1. The integrin binding sequence RLD of mouse Thy-1 has been changed to RLE abolishing the binding of integrins.