Human Recombinant CD134L (soluble)
Toimittaja: ENZO LIFE SCIENCES
ALX5220233010 ALX522023C010
ENZOALX5220233010EA 765 EUR
ENZOALX5220233010 ENZOALX522023C010
Human Recombinant CD134L (soluble)
Proteiinit
Produced in HEK 293 cells. The extracellular domain of human CD134L (OX40L) (aa 52-183) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG®-tag.