Adenosine 5'-diphosphate disodium salt (ADP disodium salt)

Toimittaja: ENZO LIFE SCIENCES

ALX480001G001 ALX480001G005
ENZOALX480001G001EA 93.5 EUR
ENZOALX480001G001 ENZOALX480001G005
Adenosine 5'-diphosphate disodium salt (ADP disodium salt)
Adenosine 5'-diphosphate disodium salt (ADP disodium salt)
Kaava: C₁₀H₁₃N₅Na₂O₁₀P₂
MDL numero: MFCD00066635
CAS-numero: 16178-48-6
EINECS: 240-314-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR