Ionomycin free acid

Toimittaja: ENZO LIFE SCIENCES

ALX450006M001 ALX450006M005
ENZOALX450006M001EA 149 EUR
ENZOALX450006M001 ENZOALX450006M005
Ionomycin free acid
Ionomycin
Kaava: C₄₁H₇₂O₉
MDL numero: MFCD06798385
CAS-numero: 56092-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR