KN-93 phosphate ≥98% (by 1H-NMR)

Toimittaja: Enzo Life Sciences

ALX430127M001 ALX430127M005
ENZOALX430127M001EA 162 EUR
ENZOALX430127M001 ENZOALX430127M005
KN-93 phosphate ≥98% (by 1H-NMR)
KN-93 phosphate

CaM kinase II inhibitor


Selective inhibitor of rat brain Ca²⁺/calmodulin-dependent protein kinase II. Inhibits glucose-induced and forskolin-stimulated insulin release from isolated pancreatic islets of Langerhans. Has no significant effect on protein kinase A (PKA) activity. Induces G1 cell cycle arrest and apoptosis in NIH 3T3 cells.

Kaava: C₂₆H₂₉ClN₂O₄S·H₃PO₄
MDL numero: MFCD03788201
CAS-numero: 1188890-41-6 | 139298-40-1 (free base)

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR