Z-IETD-Fmk
Toimittaja: ENZO LIFE SCIENCES
ALX260144R020 ALX260144R100
ENZOALX260144R020EA 275 EUR
ENZOALX260144R020 ENZOALX260144R100
Z-IETD-Fmk
Proteiinit
Caspase and granzyme inhibitor
Irreversible, cell permeable inhibitor of caspase-8 and granzyme B. Inhibits also caspase-10. The inhibitor is designed as a methyl ester to facilitate cell permeability. 