Furin convertase Substrate (fluorogenic)
Toimittaja: ENZO LIFE SCIENCES
ALX260040M001 ALX260040M005
ENZOALX260040M001EA 60.5 EUR
ENZOALX260040M001 ENZOALX260040M005
Furin convertase Substrate (fluorogenic)
Proteiinit
Fluorogenic substrate for furin/SPC1/PACE/PCSK3 and other subtilisin/kexin-like proprotein convertases such as Kex2, SPC2/PC2/PCSK2, SPC3/PCSK1, PACE4/SPC4/PCSK6, SPC5/PC4/PCSK4, SPC6/PC5/PC6/PCSK5, and SPC7/LPC/PC7/PC8/PCSK7. Can be used on live cells. Ex: 340-360 nm, Em: 440-460 nm.