Human recombinant myeloperoxidase
Toimittaja: ENZO LIFE SCIENCES
ALX200600A5025 ALX200600AU025
ENZOALX200600A5025EA 1180 EUR
ENZOALX200600A5025 ENZOALX200600AU025
Human recombinant myeloperoxidase
Proteiinit
Isolated from human leukocytes. Negative for HBsAG-, HCV- and HIV-antibodies.££Haem protein that catalyzes oxidations by H₂O₂, including MPO-chloride-mediated killing of microbes and tumor cells, inactivation of chemotactic factors, cross-linking of proteins and iodination of proteins.