Melittin
Toimittaja: ENZO LIFE SCIENCES
ALX162006M001 ALX162006M005
ENZOALX162006M001EA 101 EUR
ENZOALX162006M001 ENZOALX162006M005
Melittin
Proteiinit
PLA2 activator
Causes disruption of normal cellular activity and cell lysis. Binds calmodulin in a calcium-dependent manner. Activates phospholipase A2 (PLA2) and inhibits protein kinase C (PKC) by binding to the catalytic domain in a Mg-ATP sensitive manner. Antirheumatic.