Anti-HSP90A Rat Monoclonal Antibody
Toimittaja: ENZO LIFE SCIENCES
ADISPA840D ADISPA840F ADISPA840J
ENZOADISPA840DEA 256 EUR
ENZOADISPA840D ENZOADISPA840F ENZOADISPA840J
Anti-HSP90A Rat Monoclonal Antibody
Vasta-aineet
The Hsp90 family of heat shock proteins represents one of the most abundantly expressed and highly conserved families of cellular chaperones whose expression can be upregulated under conditions of cellular stress, and includes cytoplasmic (Hsp90-alpha/beta), ER (grp94), and mitochondrial (TRAP1) localized members. Structurally, Hsp90 is characterized by an N-terminal ATP-binding domain, a medial substrate-binding domain, and a C-terminal dimerization motif. Hsp90 dimers function in cooperation with cochaperones (e.g. Hsp40, Hsp70, Hop, p23) to stabilize a multitude of client proteinMore Product Information