Anti-HSP90 Mouse Monoclonal Antibody
Toimittaja: ENZO LIFE SCIENCES
ADISPA830D ADISPA830F ADISPA830J
ENZOADISPA830DEA 275 EUR
ENZOADISPA830D ENZOADISPA830F ENZOADISPA830J
Anti-HSP90 Mouse Monoclonal Antibody
Vasta-aineet
The Hsp90 family of heat shock proteins represents one of the most abundantly expressed and highly conserved families of cellular chaperones whose expression can be upregulated under conditions of cellular stress, and includes cytoplasmic (Hsp90-alpha/beta), ER (grp94), and mitochondrial (TRAP1) localized members. Structurally, Hsp90 is characterized by an N-terminal ATP-binding domain, a medial substrate-binding domain, and a C-terminal dimerization motif. Hsp90 dimers function in cooperation with cochaperones (e.g. Hsp40, Hsp70, Hop, p23) to stabilize a multitude of client proteinMore Product Information
Order Now
TEKNISET TIEDOT
Antigeenin symboli HSP90 HSP90
Environmentally Preferable
Vasta-aineen tyyppi Ensisijainen Ensisijainen
Klonaalisuus Monoclonal Monoclonal
Reaktiivisuus Human
Isäntä Mouse Mouse
Isotyyppi IgG1 IgG1
Western blot Yes Yes
Lomake Liquid, in PBS containing 50% glycerol and 0,09% sodium azide Liquid, in PBS containing 50% glycerol and 0,09% sodium azide
Puhdistus Protein G affinity purified Protein G affinity purified