Anti-CRP55 Rabbit Polyclonal Antibody
Toimittaja: ENZO LIFE SCIENCES
ADISPA600D ADISPA600F ADISPA600J
ENZOADISPA600DEA 241 EUR
ENZOADISPA600D ENZOADISPA600F ENZOADISPA600J
Anti-CRP55 Rabbit Polyclonal Antibody
Vasta-aineet
Calreticulin (CRT) is a multifunctional, multi-compartmental protein most abundant in the ER lumen. CRT contains the ER-retrieval sequence, KDEL, and has been best characterized as a soluble molecular chaperone of new or misfolded proteins and a Ca2+- binding protein. Both CRT and its membrane bound homolog, Calnexin (CNX) interact with proteins and glycoproteins that have monoglucosylated N-glycans. The CRT/CNX cycle promotes correct folding, inhibits aggregation of folding intermediates, blocks premature oligimerization, regulates ER degradation, and provides quality control by preventingMore Product Information