Vacuum manifold
Toimittaja: ENZO LIFE SCIENCES
ADI987500
ENZOADI987500EA 510 EUR
ENZOADI987500
Vacuum manifold
Manifoldit
The manifold is constructed of a chemical-resistant polymer and is designed to facilitate the filtration of liquid through a multi-well filter plate.