PGF2α high sensitivity ELISA kit
Toimittaja: ENZO LIFE SCIENCES
ADI930069 ADI931069
ENZOADI930069EA 517 EUR
ENZOADI930069 ENZOADI931069
PGF2α high sensitivity ELISA kit
Bio-assayt ELISA-kitit
The PGF2α high sensitivity EIA kit for the quantitative determination of PGF2α in culture supernatants, milk, plasma, serum, saliva, and urine from any species.
  • Highly sensitive quantification of PGF2α
  • Assay more samples without extraction
The PGF2α high sensitivity EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 3 hours. Absorbance is read at 405 nm.
Tilaustiedot: Kit contains DxS IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution.