Acetyltransferase activity kit
Toimittaja: ENZO LIFE SCIENCES
ADI907026
ENZOADI907026EA 669 EUR
ENZOADI907026
Acetyltransferase activity kit
Bio-assayt Solujen testausanalyysit
For the measurement of acetyl-CoA dependent acetyltransferase activity in purified in vitro samples that do not contain reductants from any species.
  • Universal - suitable for any AcetylCoA-dependent acetyltransferase
  • Robust - Z' factor = 0.87
  • Sensitive - fluorescent detection allows for both DNA and protein assays
  • Higher throughput - homogeneous mix-and-read assay
The Acetyltransferase activity kit is a homogeneous mix-and-read fluorescent assay for the determination of any acetyl-CoA dependent acetyltransferase activity. It is suitable for end-point or kinetic read options, which is ideal for determining mechanism of action, kinetics, and screening candidate compounds. The assay is amendable to HTS and miniaturization.
Tilaustiedot: Contains Black Microtiter plate, Reaction buffer, Assay buffer, Detection solution, Positive control.