Progesterone ELISA Kit
Toimittaja: ENZO LIFE SCIENCES
ADI901011
ENZOADI901011EA 1670 EUR
ENZOADI901011
Progesterone ELISA Kit
Bio-assayt ELISA-kitit
For the quantitative determination of progesterone in culture supernatants, serum, and saliva from any species. Cited sample type includes plasma.
The Progesterone EIA kit is a colorimetric competitive enzyme immunoassay kit with results in < 3 hours. Absorbance is read at 405 nm. This non-radioactive enzyme immunoassay is safe and reliable with ready-to-use liquid color-coded reagents to reduce errors.
Tilaustiedot: Contains GxM Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution, Steroid displacement reagent. Alternative size available: 89141-064 (1×96 wells).