[pSer326]HSF1 ELISA Kits
Toimittaja: ENZO LIFE SCIENCES
ADI900199
ENZOADI900199EA 700 EUR
ENZOADI900199
[pSer326]HSF1 ELISA Kits
Bio-assayt ELISA-kitit
The HSF1 EIA kit is a complete kit for the quantitative determination of HSF1 phosphorylated at Ser326 in cell lysates and tissue extracts from human and mouse origins. This is a sensitive ELISA kit for the key transcription factor for heat shock protein (chaperone) research.
  • Use less precious sample compared to Western blot
  • Time saving and convenient
  • Ready to use liquid color coded reagents and pre-coated plate
For the quantitative determination of human and mouse [pSer326]HSF1 in cell lysates and tissue.
Toimitustiedot: Kit includes antibody, assay buffer, conjugate, extraction reagent #1, microtiter plate, standard, stop solution, TMB substrate, wash buffer concentrate.