Inline gas filters, Wafergard® III NF Mini and Mini XL
Toimittaja: ENTEGRIS
WG3NS2RR2 WG3NS2RR4 WG3NS1RR2
ENTEWG3NS2RR2EA 0 EUR
ENTEWG3NS2RR2 ENTEWG3NS2RR4 ENTEWG3NS1RR2
Inline gas filters, Wafergard® III NF Mini and Mini XL
Suodattimet Kaasusuodattimet
The patented nickel filter membrane of these inline gas filters offers superior corrosion resistance and excellent compatibility with inert and reactive gases. Not recommended for use with CO, O₃ and low-level hydride dopant gases.
  • Superior particulate filtration for ultrapure gas system filtration
  • Housing assembly eliminates outgassing by minimising weld area
  • Eliminates outgassing, improves corrosion resistance and provides excellent chemical compatibility
Filter element: patented, sintered nickel membrane; housing: electropolished VAR 316L stainlessMore Product Information