Quick Connect System Sentry® QCII
Toimittaja: ENTEGRIS
DIP-29-02-100 DHT-00-09-000 DHT-00-02-000 DHT-00-06-000 DHT-00-08-000 DHT-00-01-000 DHT-00-05-000 DHT-00-03-000 DIT-01-01-000 DIP-29-09-006 DHT-00-03-004 DHT-00-07-000 DHT-00-10-000 DHT-00-04-000 DHT-00-12-000 DHT-00-13-000 DIP-29-02-104 DIP-29-03-000 DIP-29-03-004 DIP-29-03-100 DIP-29-05-000 DIP-29-05-100 DIP-29-05-109 DIP-29-06-000 DIP-29-06-104 DIP-29-08-100 DIP-29-08-104 DIP-29-09-000 DIP-29-09-104 DIP-29-09-106 DIP-53-02-100 DIP-53-03-000 DIP-53-03-003 DIP-53-03-004 DIP-53-05-103 DIP-53-05-104 DIP-53-05-109 DIP-53-06-100 DIP-53-06-104 DIP-53-08-100 DIP-53-08-104 DIP-53-13-100 DIT-01-01-004 DIT-01-01-100 DIT-01-02-100 DIT-01-05-000 DIT-01-06-100 DIT-01-08-000 DIT-29-01-000 DIT-29-02-000 DIT-29-03-000 DIT-29-03-003 DIT-29-05-000 DIT-29-06-000 DIT-29-06-105 DIT-29-07-000 DIT-29-08-000 DIT-29-09-000 DIT-29-09-006 DIT-01-09-100
ENTEDIP-29-02-100EA 0 EUR
ENTEDIP-29-02-100 ENTEDHT-00-09-000 ENTEDHT-00-02-000 ENTEDHT-00-06-000 ENTEDHT-00-08-000 ENTEDHT-00-01-000 ENTEDHT-00-05-000 ENTEDHT-00-03-000 ENTEDIT-01-01-000 ENTEDIP-29-09-006 ENTEDHT-00-03-004 ENTEDHT-00-07-000 ENTEDHT-00-10-000 ENTEDHT-00-04-000 ENTEDHT-00-12-000 ENTEDHT-00-13-000 ENTEDIP-29-02-104 ENTEDIP-29-03-000 ENTEDIP-29-03-004 ENTEDIP-29-03-100 ENTEDIP-29-05-000 ENTEDIP-29-05-100 ENTEDIP-29-05-109 ENTEDIP-29-06-000 ENTEDIP-29-06-104 ENTEDIP-29-08-100 ENTEDIP-29-08-104 ENTEDIP-29-09-000 ENTEDIP-29-09-104 ENTEDIP-29-09-106 ENTEDIP-53-02-100 ENTEDIP-53-03-000 ENTEDIP-53-03-003 ENTEDIP-53-03-004 ENTEDIP-53-05-103 ENTEDIP-53-05-104 ENTEDIP-53-05-109 ENTEDIP-53-06-100 ENTEDIP-53-06-104 ENTEDIP-53-08-100 ENTEDIP-53-08-104 ENTEDIP-53-13-100 ENTEDIT-01-01-004 ENTEDIT-01-01-100 ENTEDIT-01-02-100 ENTEDIT-01-05-000 ENTEDIT-01-06-100 ENTEDIT-01-08-000 ENTEDIT-29-01-000 ENTEDIT-29-02-000 ENTEDIT-29-03-000 ENTEDIT-29-03-003 ENTEDIT-29-05-000 ENTEDIT-29-06-000 ENTEDIT-29-06-105 ENTEDIT-29-07-000 ENTEDIT-29-08-000 ENTEDIT-29-09-000 ENTEDIT-29-09-006 ENTEDIT-01-09-100
Quick Connect System Sentry® QCII
Kiinnitykset ja liittimet
The safe and convenient method of interfacing reusable FluoroPure® chemical containers with bulk delivery systems. The patented design is key coded to assure against unsafe chemical mixing while ensuring dedicated clean service. One connection provides dispensing, venting and level detection.
  • A radial seal design provides superior leak-free performance even after multiple make-and-break connections, while minimising o-ring wear
  • Dispense head has an independently rotated key code ring that allows quick key tab alignment without having to twist the