Galtek® Check Valves, ³/₄" Orifice, O-ring Free Design E
Toimittaja: ENTEGRIS
CKM12-12F-12F CKM12-12K-12K
ENTECKM12-12F-12FEA 0 EUR
ENTECKM12-12F-12F ENTECKM12-12K-12K
Galtek® Check Valves, ³/₄" Orifice, O-ring Free Design E
Venttiilit Tarkista venttiilit
Corrosion-resistant Galtek® check valves reduce contamination potential in your high-purity chemical applications and provide clean delivery of sophisticated chemicals.
  • O-ring free design reduces potential contamination
  • Simple, clean, compact valve provides high flow chemical delivery
  • No metal parts - will not corrode or contribute to contamination
  • Poppet design allows a high flow through rate and maximizes rinsability
An O-ring-free design provides a simple, clean, compact valve with a high flow rate and increased reliability. All wetted parts areMore Product Information