Lisävarusteet, Wafer carrier handles, squeeze style
Toimittaja: ENTEGRIS
A72-40-03-0215 A72-40-06-0215
ENTEA72-40-03-0215EA 0 EUR
ENTEA72-40-03-0215 ENTEA72-40-06-0215
Lisävarusteet, Wafer carrier handles, squeeze style
Piikiekkojen käsittely Kasetti
These wafer warrier handles are designed for extended lead times apply.
  • Attaches to the notches on both end walls of carrier
  • Compatible with all 'M' style wafer carriers