Fluid aspirator, Galtek®
Toimittaja: ENTEGRIS
226-7 226-5 226-1 226-2 226-3 226-4 226-6
ENTE226-7EA 0 EUR
ENTE226-7 ENTE226-5 ENTE226-1 ENTE226-2 ENTE226-3 ENTE226-4 ENTE226-6
Fluid aspirator, Galtek®
Pumput Imujärjestelmä
Galtek® aspirator is ideal for the extraction and/or dilution of fluids from chemical tanks.
  • Made from PFA for long lasting corrosion resistance
  • Requires only water or air to operate