Galtek® solenoid operated diaphragm valves
Toimittaja: ENTEGRIS
203-1414-215 203-3414-215
ENTE203-1414-215EA 0 EUR
ENTE203-1414-215 ENTE203-3414-215
Galtek® solenoid operated diaphragm valves
Venttiilit Vakuumiventtiilit
Two- or three-way diaphragm valves, ¹/₄" orifice, normally closed.
  • All wetted parts are PFA and Kalrez® for high purity and corrosion resistance
  • Wide range of voltages to select from
  • Powerful but compact coil design