Test sieves, 200×50 mm
Toimittaja: ENDECOTTS
200SIW.020 200SIW.025 200SIW.032 200SIW.036 200SIW.038 200SIW.040 200SIW.050 200SIW.053 200SIW.056 200SIW.063 200SIW.071 200SIW.075 200SIW.080 200SIW.090 200SIW.100 200SIW.106 200SIW.125 200SIW.140 200SIW.150 200SIW.160 200SIW.180 200SIW.200 200SIW.212 200SIW.224 200SIW.250 200SIW.280 200SIW.300 200SIW.315 200SIW.355 200SIW.400 200SIW.425 200SIW.450 200SIW.500 200SIW.560 200SIW.600 200SIW.630 200SIW.800 200SIW.850 200SIW.900 200SIW1.00 200SIW1.12 200SIW1.18 200SIW1.25 200SIW1.40 200SIW1.60 200SIW1.70 200SIW1.80 200SIW10.0 200SIW100 200SIW106 200SIW11.2 200SIW112 200SIW125 200SIW13.2 200SIW14.0 200SIW16.0 200SIW18.0 200SIW19.0 200SIW2.00 200SIW2.24 200SIW2.36 200SIW2.50 200SIW2.80 200SIW20.0 200SIW22.4 200SIW25.0 200SIW26.5 200SIW28.0 200SIW3.15 200SIW3.35 200SIW3.55 200SIW31.5 200SIW35.5 200SIW37.5 200SIW4.00 200SIW4.50 200SIW4.75 200SIW40.0 200SIW45.0 200SIW5.00 200SIW5.60 200SIW50.0 200SIW53.0 200SIW56.0 200SIW6.30 200SIW6.70 200SIW63.0 200SIW7.10 200SIW75.0 200SIW8.00 200SIW80.0 200SIW9.00 200SIW9.50 200SIW90.0 200SIW12.5 200SIW.710 200SIW.112 200SIW.045 200SIW71.0
ENDE200SIW.020EA 1020 EUR
ENDE200SIW.020 ENDE200SIW.025 ENDE200SIW.032 ENDE200SIW.036 510-0274 ENDE200SIW.040 ENDE200SIW.050 510-0276 ENDE200SIW.056 510-0277 ENDE200SIW.071 510-0278 ENDE200SIW.080 510-0279 ENDE200SIW.100 510-0280 510-0281 ENDE200SIW.140 510-0282 ENDE200SIW.160 510-0283 ENDE200SIW.200 510-0284 ENDE200SIW.224 510-0285 ENDE200SIW.280 510-0286 ENDE200SIW.315 510-0372 ENDE200SIW.400 510-0287 ENDE200SIW.450 510-0288 ENDE200SIW.560 510-0307 ENDE200SIW.630 ENDE200SIW.800 510-0290 ENDE200SIW.900 510-0291 ENDE200SIW1.12 510-0292 ENDE200SIW1.25 510-0293 ENDE200SIW1.60 510-0294 ENDE200SIW1.80 ENDE200SIW10.0 ENDE200SIW100 ENDE200SIW106 510-0295 ENDE200SIW112 ENDE200SIW125 510-0296 ENDE200SIW14.0 510-0297 ENDE200SIW18.0 ENDE200SIW19.0 510-0298 ENDE200SIW2.24 510-0299 ENDE200SIW2.50 510-0300 ENDE200SIW20.0 ENDE200SIW22.4 ENDE200SIW25.0 ENDE200SIW26.5 ENDE200SIW28.0 ENDE200SIW3.15 510-0301 ENDE200SIW3.55 ENDE200SIW31.5 ENDE200SIW35.5 ENDE200SIW37.5 510-0303 ENDE200SIW4.50 510-0304 ENDE200SIW40.0 ENDE200SIW45.0 ENDE200SIW5.00 510-0305 ENDE200SIW50.0 ENDE200SIW53.0 ENDE200SIW56.0 ENDE200SIW6.30 510-0306 ENDE200SIW63.0 ENDE200SIW7.10 ENDE200SIW75.0 510-0308 ENDE200SIW80.0 ENDE200SIW9.00 510-0309 ENDE200SIW90.0 ENDE200SIW12.5 510-0289 ENDE200SIW.112 510-0275 ENDE200SIW71.0
Test sieves, 200×50 mm
Seulat Testisiivilät
Stainless steel woven wire mesh test sieves are ideal for all types of laboratory sampling and particle size analysis.
For more size options please contact your local Avantor office.
Sertifikaatit: ISO3310-1:2000, BS410-1:2000; DIN-ISO3310-1; NF ISO3310-1:2000
Toimitustiedot: Each sieve is supplied together with a certificate of examination and carries the BS 410 kite mark.