Elma Lab Clean Cleaning Concentrates for Laboratory
Toimittaja: ELMA SCHMIDBAUER
8000180 8000181 8000182 8000077 8000076 8000193 8000078 8000085 8000086 8000087 8000088 8000183 8000190 8000191 8000192 8000075
ELMA8000180EA 28.8 EUR
ELMA8000180 ELMA8000181 ELMA8000182 ELMA8000077 ELMA8000076 ELMA8000193 ELMA8000078 ELMA8000085 ELMA8000086 ELMA8000087 ELMA8000088 ELMA8000183 ELMA8000190 ELMA8000191 ELMA8000192 ELMA8000075
Elma Lab Clean Cleaning Concentrates for Laboratory
Puhdistusaineet Pesuaineet ja pintojen puhdistusaineet
Laboratory cleaning agents designed to meet requirements for instrumentation and glassware in analysis laboratories. All Elma Lab Clean cleaning agents are formulated for use with ultrasonic intensive cleaning devices.
These products are also suitable for laboratory rinsing machines, as they are highly effective without foaming.