Cleaning agents, elma tec clean
Toimittaja: ELMA SCHMIDBAUER
8000170 8000172 8000152 8000102 8000122 8000150 8000112 8000132
ELMA8000170EA 380 EUR
ELMA8000170 ELMA8000172 ELMA8000152 ELMA8000102 ELMA8000122 ELMA8000150 ELMA8000112 ELMA8000132
Cleaning agents, elma tec clean
Puhdistusaineet Pesuaineet ja pintojen puhdistusaineet
Powerful cleaners for ultrasonic baths suitable for many industrial cleaning tasks
  • prolongs the service life of the ultrasonic unit
  • Highly efficient concentrate
  • Long shelf life