Perfluoropolyether oil for rotary vacuum pumps, Fomblin® YVAC
Toimittaja: EDWARDS HIGH VACUUM METTLER
H11301019
EDWAH11301019EA 970 EUR
EDWAH11301019
Perfluoropolyether oil for rotary vacuum pumps, Fomblin® YVAC
Rasvat
Fomblin® perfluoropolyether oil for use in oil sealed rotary vacuum pumps and are practically a direct replacement for mineral oil in terms of viscosity and vapour pressure. Also it contains Drynert fluid which contains anti-rust and anti-wear additives soluble in Fomblin®. These additives cover metallic surfaces with a protective, corrosion resistant film.
It is widely used for semiconductors, chemical pumping, oxygen pumping, radioactive and metallisation.