Rotary vane pumps, RV ATEX
Toimittaja: EDWARDS HIGH VACUUM METTLER
A65518989 A65518991
EDWAA65518989EA 0 EUR
EDWAA65518989 EDWAA65518991
Rotary vane pumps, RV ATEX
Pumput Siipipyörävakuumipumput
ATEX RV pumps have an external ATEX classification of Ex II 2G IIC T4 or Ex II 2G IIB T4 and internal ATEX classification Ex II 3G c IIB T4. They are suitable for use in potentially explosive atmospheres such as hazardous area pumping. They are supplied with the gas ballast port which must be fitted with an inert gas purge while pumping flammable gasses or vapours to dilute such gases or vapours below the lower explosive limit.
  • ATEX rated spot voltage motor
  • Suitable for high throughput and high vacuum applications
  • Fast acting inlet valve for system