Ready-to-Load™ kits, principles and practice of agarose gel electrophoresis
Toimittaja: EDVOTEK
101-C 101
EDVO101-CEA 164 EUR
EDVO101-C EDVO101
Ready-to-Load™ kits, principles and practice of agarose gel electrophoresis
Opetusmateriaalit Biology Educational Materials Genetics Learning Activities
Ready-to-Load™ kit to show how electrophoresis separates molecules on the basis of size and charge.
  • For 6 gels
  • Complete in 45 minutes
Also required: Electrophoresis tank, power supply, adjustable micropipettes (5 to 50 µl) with tips, balance and microwave or hot plate.
Toimitustiedot: Kit includes instructions, Ready-to-Load™ dye samples, UltraSpec-Agarose™ powder, practice gel loading solution, electrophoresis buffer, calibrated micropipette, 100 ml graduated cylinder, and microtipped transfer pipettes.