VWR®, Bovine Ribonuclease A (from Pancreas)
Toimittaja: VWR Chemicals
E866-1MLEA 68.2 EUR
E866-1ML E866-5ML
VWR®, Bovine Ribonuclease A (from Pancreas)
Entsyymit
Ribonuclease A is an endoribonuclease that efficiently hydrolyses RNA from tissue or bacterial cell cultures.
  • Useful during DNA purification
  • High purity
  • Available in a ready-to-use solution
RNase A is used for a variety of molecular biology applications, most commonly for the hydrolysis of RNA that contaminates DNA preparations. This enzyme is used during plasmid purification without nicking or degrading plasmid.