Dekstraanisulfaatin natriumsuola, M.W. ~ 500,000 50% for biotechnology

Toimittaja: VWR Chemicals
Varoitus

E516-100MLEA 322 EUR
E516-100ML
Dekstraanisulfaatin natriumsuola, M.W. ~ 500,000 50% for biotechnology
Dekstraanisulfaatin natriumsuola

Used for the detection of DNA or RNA in nucleic acid hybridisation procedures. It effectively “excludes” probe from solution, accelerating the rate of annealing.

Kaava: (C₆H₇Na₃O₁₄S₃)n
MDL numero: MFCD00081551
CAS-numero: 9011-18-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae52589f20d0da38bdceca63c873f7fba4580dd4&vid=bf56737a35089faf2295e8fcb2d8a1ea654b245a&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ddbddce328004684d12a8dc9f1b65d847c433847&vid=f9e15aedab27c83fb2129e9d24b773e8c42b2a7c&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

APHA (1:10) Tan to yellow haze-free liquid
DNase none detected
RNase none detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR